Learn More
Meso Scale Discovery U-PLEX NHP CTACK ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K156VDK4
Encompass
The Meso Scale Discovery U-PLEX NHP CTACK Assay Kit is a multiplex immunoassay that measures CTACK (Cutaneous T-Cell Attracting Chemokine) levels in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators and diluent This allows for the accurate quantitation of CTACK levels with high sensitivity and a wide dynamic range The assay takes approximately 3 5 hours to complete and requires a MSD instrument for analysis
Catalog No. 50-000-80037
Add to Cart
Request a Quote