Learn More
Meso Scale Discovery U-PLEX NHP ENA-78 ASY384-well
SDP
Supplier:  Meso Scale Discovery K256VEK2
Encompass
The U-PLEX NHP ENA 7-8-384 Well Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of autoantibodies to extractable nuclear antigens in human serum plasma and other biological fluids This 384-well multiplex assay kit allows simultaneous detection of autoantibodies to 7 antigens including RNP Sm SS-A SS-B Scl-70 Jo-1 and CENP-B in a single small sample volume The kit includes a 384-well standard microplate coated with antigens detection antibodies read buffer wash buffer calibrators and controls This assay provides high sensitivity requires small sample volume and has a quick turnaround time
Catalog No. 50-000-79200
Only null left
Add to Cart
Request a Quote