Learn More
Meso Scale Discovery U-PLEX NHP Eotaxin ASY384-well
SDP
Supplier:  Meso Scale Discovery K256UDK2
Encompass
The Meso Scale Discovery U-PLEX NHP Eotaxin 384-well Assay Kit is a multiplex immunoassay that measures Eotaxin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput screening This quantitative assay has high sensitivity wide dynamic range and requires low sample volume It includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels and calibrators and controls to generate a standard curve for quantitation The ready-to-use reagents provide convenience and consistent results
Catalog No. 50-000-81342
Add to Cart
Request a Quote