Learn More
Meso Scale Discovery U-PLEX NHP FLT3L ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K156XFK4
Encompass
The Meso Scale Discovery U-PLEX NHP FLT3L Assay is an electrochemiluminescent immunoassay for measuring human FLT3 ligand in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive results in 3 5 hours The detection limit is 0 11 pg/mL and the quantification limit is 2500 pg/mL It requires 50 L of sample and shows no cross-reactivity with other cytokines The kit includes a 96-well plate antibodies buffers reagents calibrator and controls It is for research use only
Catalog No. 50-000-80734
Add to Cart
Request a Quote