Learn More
Meso Scale Discovery U-PLEX NHP FLT3L ASYQP
SDP
Supplier:  Meso Scale Discovery K156XFK21
Encompass
The U-PLEX NHP FLT3L Assay (K156XFK) is a specialized immunoassay designed to measure FLT3 ligand levels in non-human primate samples This electrochemiluminescent detection platform offers high sensitivity and specificity for research applications studying hematopoietic growth factors The assay utilizes MSD s U-PLEX technology for reliable quantification of FLT3L protein concentrations in biological samples supporting preclinical research and drug development studies involving immune system regulation and stem cell biology
Catalog No. 50-000-80915