Learn More
Meso Scale Discovery U-PLEX NHP G-CSF ASY
SDP
Supplier:  Meso Scale Discovery K156VGK2
Encompass
The Meso Scale Discovery U-PLEX NHP CSF Assay is a multiplex immunoassay panel measuring NfL t-tau p-tau A 42 A 40 YKL-40 neurogranin and sTREM2 in human CSF Utilizing MSD s electrochemiluminescent detection it provides quantitative results in under 5 hours from a small sample Intended for investigational use by biopharmaceutical companies and researchers it aids in understanding neurological diseases and assessing therapeutics The assay is highly sensitive reproducible and requires minimal hands-on time
Catalog No. 50-000-80065