Learn More
Meso Scale Discovery U-PLEX NHP GM-CSF ASY
SDP
Supplier:  Meso Scale Discovery K156UMK1
Encompass
The Meso Scale Discovery U-PLEX NHP GM-CSF Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of natural human pro-granulocyte macrophage colony-stimulating factor (GM-CSF) in cell culture supernatants serum plasma and other biological matrices This assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The assay has high sensitivity and wide dynamic range and requires only 50 L sample volume per well The kit includes a plate coated with capture antibodies detection antibodies calibrators and diluents needed to perform the assay
Catalog No. 50-000-80059