Learn More
Meso Scale Discovery U-PLEX NHP GRO- ASY
SDP
Supplier:  Meso Scale Discovery K156UXK2
Encompass
The Meso Scale Discovery U-PLEX NHP GRO-alpha Assay is a multiplex immunoassay kit for quantifying human GRO-alpha in serum plasma and cell culture supernatants It uses electrochemiluminescent detection for high sensitivity and precision The assay requires 50 L or less sample volume and takes about 3 5 hours It includes a 96-well plate detection antibodies calibrators controls buffers and seals Sensitivity is 0 22 pg/mL with an upper quantification limit of 10 000 pg/mL
Catalog No. 50-000-80679