Learn More
Meso Scale Discovery U-PLEX NHP I-309 ASY
SDP
Supplier:  Meso Scale Discovery K156UYK2
Encompass
The U-PLEX NHP-I 309 Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that measures 9 biomarkers related to neuroinflammation and neurodegeneration This assay uses electrochemiluminescence detection technology and is designed for use on Meso Scale Diagnostics instrumentation The panel includes assays for the following analytes A 40 A 42 -Synuclein T-tau P-tau NFL GFAP sTREM2 and YKL-40 This assay allows simultaneous quantification of these biomarkers from a small sample volume It is intended for research use only
Catalog No. 50-000-80140