Learn More
Meso Scale Discovery U-PLEX NHP IL-10 ASY
SDP
Supplier:  Meso Scale Discovery K156TZK2
Encompass
The Meso Scale Discovery U-PLEX NHP IL-10 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 10 (IL-10) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 04 pg/mL The assay can be multiplexed with other U-PLEX biomarkers for efficient and customizable biomarker profiling
Catalog No. 50-000-81319