Learn More
Meso Scale Discovery U-PLEX NHP IL-12p70 ASY
SDP
Supplier:  Meso Scale Discovery K156UAK1
Encompass
The Meso Scale Discovery U-PLEX NHP IL-12p70 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human IL-12p70 in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 0 6 pg/mL and the assay range is 0 6-2500 pg/mL
Catalog No. 50-000-79933