Learn More
Meso Scale Discovery U-PLEX NHP IL-15 ASY
SDP
Supplier:  Meso Scale Discovery K156URK2
Encompass
The Meso Scale Discovery U-PLEX NHP IL-15 Assay is an electrochemiluminescent immunoassay for measuring IL-15 in human cynomolgus monkey or rhesus macaque samples It uses MSD s U-PLEX technology for accurate quantitation with a range of 19 5 to 10 000 pg/mL The assay requires 25 L of sample and shows high specificity without cross-reactivity The kit includes a 96-well plate with IL-15 capture antibody detection antibody buffers and a lyophilized calibrator
Catalog No. 50-000-80120