Learn More
Meso Scale Discovery U-PLEX NHP IL-17A ASY384-well
SDP
Supplier:  Meso Scale Discovery K256UTK2
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17A 384-Well Assay Kit is a multiplex immunoassay that measures natural human IL-17A levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid time-to-result The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels and calibrators and controls needed to generate a standard curve and validate assay performance It allows high throughput analysis of IL-17A an important proinflammatory cytokine with a testing volume of 25 L per well
Catalog No. 50-000-80225
Only null left
Add to Cart
Request a Quote