Learn More
Meso Scale Discovery U-PLEX NHP IL-17B ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K156XNK4
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17B Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of IL-17B in non-human primate (NHP) samples such as serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The assay has high sensitivity wide dynamic range and requires low sample volume This product can aid research on the role of IL-17B in inflammation and autoimmune diseases using NHP models With minimal hands-on time required this assay provides a simple and rapid workflow for analyzing IL-17B levels in NHP studies
Catalog No. 50-000-78618
Add to Cart
Request a Quote