Learn More
Meso Scale Discovery U-PLEX NHP IL-17B ASY
SDP
Supplier:  Meso Scale Discovery K156XNK2
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17B Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of IL-17B in non-human primate serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance Turnaround time is 3 5 hours for up to 80 samples This assay allows reliable reproducible quantification of IL-17B
Catalog No. 50-000-80739