Learn More
Meso Scale Discovery U-PLEX NHP IL-17B ASYQP
SDP
Supplier:  Meso Scale Discovery K156XNK21
Encompass
The U-PLEX NHP IL-17B Assay (K156XNK) is a specialized immunoassay kit designed for quantitative measurement of Interleukin-17B in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for cytokine detection in research applications The kit provides researchers with validated reagents and protocols for studying IL-17B levels in preclinical studies involving non-human primate models
Catalog No. 50-000-80875