Learn More
Meso Scale Discovery U-PLEX NHP IL-17C ASY
SDP
Supplier:  Meso Scale Discovery K156WJK2
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17C Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of IL-17C in non-human primate (NHP) samples such as serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This assay allows for efficient reproducible quantification of NHP IL-17C levels
Catalog No. 50-000-79907