Learn More
Meso Scale Discovery U-PLEX NHP IL-17D ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K156XOK4
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17D Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of IL-17D in non-human primate (NHP) samples such as serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The assay has high sensitivity and wide dynamic range Each kit provides reagents for one 96-well plate and can test 40 samples in duplicate Assay duration is approximately 3 hours This product enables reliable high-throughput analysis of IL-17D which plays a role in inflammatory and autoimmune responses
Catalog No. 50-000-78635
Only null left
Add to Cart
Request a Quote