Learn More
Meso Scale Discovery U-PLEX NHP IL-17F ASY
SDP
Supplier:  Meso Scale Discovery K156WAK2
Encompass
The Meso Scale Discovery U-PLEX NHP IL-17F Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of human interleukin 17F (IL-17F) in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a wide dynamic range and requires low sample volume The kit includes a plate coated with capture antibodies detection antibodies calibrators controls 10X read buffer 10X wash buffer and substrate The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL for IL-17F
Catalog No. 50-000-79782