Learn More
Meso Scale Discovery U-PLEX NHP IL-22 ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K156WIK4
Encompass
The Meso Scale Discovery U-PLEX NHP IL-22 Assay Kit is a sensitive immunoassay for measuring human IL-22 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibody detection antibody read buffer calibrator diluent wash buffer and sample diluent Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL enabling accurate IL-22 quantification in human samples
Catalog No. 50-000-80068
Add to Cart
Request a Quote