Learn More
Meso Scale Discovery U-PLEX NHP IL-5 ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K156UOK24
Encompass
The U-PLEX NHP IL-5 Assay (K156UOK) is a specialized immunoassay kit designed for quantitative measurement of interleukin-5 (IL-5) in non-human primate samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity and specificity for cytokine detection in research applications The kit provides researchers with validated reagents and protocols for accurate IL-5 quantification in serum plasma and cell culture supernatants from non-human primate species supporting immunology and inflammation research studies
Catalog No. 50-000-80677
Only null left
Add to Cart
Request a Quote