Learn More
Meso Scale Discovery U-PLEX NHP IL-6 ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K156TXK4
Encompass
The Meso Scale Discovery U-PLEX NHP IL-6 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 6 (IL-6) in serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a wide dynamic range The assay has high specificity for IL-6 with no cross-reactivity to other cytokines It requires low sample volume and has a rapid procedure time of 3 5 hours The lower limit of detection is 0 10 pg/mL This assay allows for the precise quantification of IL-6 levels in human samples
Catalog No. 50-000-79912
Add to Cart
Request a Quote