Learn More
Meso Scale Discovery U-PLEX NHP IL-6 ASY
SDP
Supplier:  Meso Scale Discovery K156TXK1
Encompass
The Meso Scale Discovery U-PLEX NHP IL-6 Assay Kit is an electrochemiluminescent immunoassay for measuring human interleukin 6 (IL-6) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology offering a detection range of 0 06-1500 pg/mL and a lower limit of 0 03 pg/mL The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are read on MSD s SECTOR Imager and analyzed with Discovery Workbench software providing a rapid high-throughput method for IL-6 measurement
Catalog No. 50-000-81138