Learn More
Meso Scale Discovery U-PLEX NHP IL-7 ASY384-well
SDP
Supplier:  Meso Scale Discovery K256UPK2
Encompass
The Meso Scale Discovery U-PLEX NHP IL-7 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The assay has a sensitivity of 0 11 pg/mL and a 7-point standard curve from 0 3 to 2000 pg/mL It uses MSD GOLD SULFO-TAG labels and carbon electrodes in microplates for high sensitivity and low background The kit includes a plate coated with IL-7 capture antibody detection antibody read buffer wash buffer blocker solutions calibration solutions and plate seals The assay takes 3 5 hours total time with 2 incubation steps
Catalog No. 50-000-79343
Only null left
Add to Cart
Request a Quote