Learn More
Meso Scale Discovery U-PLEX NHP IL-8 ASY384-well
SDP
Supplier:  Meso Scale Discovery K256TYK2
Encompass
The Meso Scale Discovery U-PLEX NHP IL-8 384-Well Assay Kit is a multiplex immunoassay that measures human IL-8 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluent It allows simultaneous measurement of IL-8 across two logs of range from 0 06 to 1500 pg/mL The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-78976
Only null left
Add to Cart
Request a Quote