Learn More
Meso Scale Discovery U-PLEX NHP IL-9 ASY384-well
SDP
Supplier:  Meso Scale Discovery K256XKK2
Encompass
The Meso Scale Discovery U-PLEX NHP IL-9 384-Well Assay Kit is a 384-well multiplex assay that measures human IL-9 in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 384-well MULTI-ARRAY Plate Calibrator Blend Diluent 35 Read Buffer T (4X) and ancillary reagents and materials to run the assay
Catalog No. 50-000-81767
Add to Cart
Request a Quote