Learn More
Meso Scale Discovery U-PLEX NHP IL-9 ASYQP
SDP
Supplier:  Meso Scale Discovery K156XKK22
Encompass
The U-PLEX NHP IL-9 Assay (K156XKK) is a specialized immunoassay designed for measuring interleukin-9 levels in non-human primate samples This research tool utilizes Meso Scale Discovery s electrochemiluminescence detection technology to provide sensitive and accurate quantification of IL-9 a cytokine involved in immune responses and inflammatory processes The assay is specifically validated for use with serum plasma and cell culture supernatants from non-human primate species making it valuable for preclinical research and drug development studies requiring precise IL-9 measurement
Catalog No. 50-000-80737