Learn More
Meso Scale Discovery U-PLEX NHP M-CSF ASY
SDP
Supplier:  Meso Scale Discovery K156XRK2
Encompass
The U-PLEX NHP CSF Assay from Meso Scale Diagnostics is a multiplex immunoassay panel measuring neurofilament light chain total tau phosphorylated tau amyloid beta 42 amyloid beta 40 YKL-40 neurogranin and GFAP in human cerebrospinal fluid Utilizing electrochemiluminescence detection it provides quantitative results in under 3 hours with a small sample volume Intended for investigational use it aids biopharmaceutical companies and researchers in understanding neurological diseases offering sensitive biomarker measurements for neurodegenerative research insights
Catalog No. 50-000-80740