Learn More
Meso Scale Discovery U-PLEX NHP MCP-1 ASY384-well
SDP
Supplier:  Meso Scale Discovery K256UGK2
Encompass
The Meso Scale Discovery U-PLEX NHP MCP-1 384-well Assay Kit is a multiplex immunoassay for measuring human MCP-1 in various samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible quantification with just 25 L of sample The kit includes a 384-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on MSD s SECTOR Imager systems enabling high-throughput MCP-1 profiling in research
Catalog No. 50-000-80203
Add to Cart
Request a Quote