Learn More
Meso Scale Discovery U-PLEX NHP MCP-1 ASYQP
SDP
Supplier:  Meso Scale Discovery K156UGK22
Encompass
The U-PLEX NHP MCP-1 Assay (K156UGK) is a specialized immunoassay kit designed for quantitative measurement of Monocyte Chemoattractant Protein-1 (MCP-1) in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit provides researchers with validated reagents and protocols for detecting MCP-1 a key inflammatory biomarker involved in immune cell recruitment and inflammatory responses in preclinical studies
Catalog No. 50-000-80647