Learn More
Meso Scale Discovery U-PLEX NHP MDC ASYQP
SDP
Supplier:  Meso Scale Discovery K156UIK22
Encompass
The U-PLEX NHP MDC Assay (K156UIK) is a specialized immunoassay kit designed for measuring macrophage-derived chemokine (MDC/CCL22) levels in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for research applications The kit provides researchers with validated reagents and protocols for quantitative detection of MDC a key biomarker involved in immune responses and inflammatory processes in preclinical studies using non-human primate models
Catalog No. 50-000-80644