Learn More
Meso Scale Discovery U-PLEX NHP MIF ASYQP
SDP
Supplier:  Meso Scale Discovery K156XJK22
Encompass
The U-PLEX NHP MIF Assay (K156XJK) is a specialized immunoassay kit designed for measuring Macrophage Inhibitory Factor (MIF) levels in non-human primate samples This electrochemiluminescent detection platform offers high sensitivity and specificity for research applications studying inflammatory responses and immune system function in preclinical models The assay provides reliable quantitative results for biomarker analysis in pharmaceutical and academic research settings
Catalog No. 50-000-78374