Learn More
Meso Scale Discovery U-PLEX NHP MIP-1B ASY
SDP
Supplier:  Meso Scale Discovery K156UKK2
Encompass
The Meso Scale Discovery U-PLEX NHP MIP-1 Assay is a multiplex immunoassay kit for measuring Macrophage Inflammatory Protein 1-beta (MIP-1 ) in non-human primate samples like serum plasma and culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls for standard curve generation and assay validation enabling efficient quantification of MIP-1
Catalog No. 50-000-79749