Learn More
Meso Scale Discovery U-PLEX NHP MIP-1B ASY384-well
SDP
Supplier:  Meso Scale Discovery K256UKK2
Encompass
The Meso Scale Discovery U-PLEX NHP MIP-1beta 384-well Assay Kit is a multiplex immunoassay that measures human MIP-1beta in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This 384-well plate assay allows for efficient and high-throughput screening The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Sample Diluent and calibrators to generate a standard curve It can detect MIP-1beta concentrations as low as 17 pg/mL with an upper limit of 100 000 pg/mL
Catalog No. 50-000-81344
Add to Cart
Request a Quote