Learn More
Meso Scale Discovery U-PLEX NHP MIP-5 ASY
SDP
Supplier:  Meso Scale Discovery K156XSK1
Encompass
The Meso Scale Discovery U-PLEX NHP MIP 5 Assay is a multiplex immunoassay panel measuring 5 biomarkers in human serum plasma and CSF It uses electrochemiluminescence detection for quantitative results of A 42 t-tau p-tau -synuclein and NFL with high sensitivity from a small sample This panel assesses pathology and disease progression in Alzheimer s Parkinson s and related disorders The kit includes a 96-well plate detection antibodies calibrators controls read buffer sample diluent and plate seals for one plate
Catalog No. 50-000-80900