Learn More
Meso Scale Discovery U-PLEX NHP MIP-5 ASY384-well
SDP
Supplier:  Meso Scale Discovery K256XSK2
Encompass
The U-PLEX NHP MIP 5-Plex 384-well Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay panel for simultaneous quantitative measurement of 5 neurodegenerative biomarkers (A 40 A 42 p-tau t-tau and -synuclein) in a 384-well plate format using electrochemiluminescent detection It offers a sensitive reproducible method for analyzing cerebrospinal fluid and blood plasma aiding research and drug development for Alzheimer s Parkinson s and other neurodegenerative disorders The kit includes reagents and plates for up to 384 samples
Catalog No. 50-000-81772
Add to Cart
Request a Quote