Learn More
Meso Scale Discovery U-PLEX NHP SDF-1 ASY
SDP
Supplier:  Meso Scale Discovery K156VBK1
Encompass
The U-PLEX NHP SDF-1 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay kit that allows for the quantitative measurement of human Stromal Cell-Derived Factor 1 alpha (SDF-1 ) in serum plasma and cell culture supernatants The assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrators and controls The assay has a typical sensitivity of 0 64 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-79741