Learn More
Meso Scale Discovery U-PLEX NHP SDF-1 ASY
SDP
Supplier:  Meso Scale Discovery K156VBK2
Encompass
The U-PLEX NHP SDF-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human SDF-1 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive quantification with minimal sample volume The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are analyzed on the MESO QuickPlex SQ 120 instrument aiding research in immunology oncology and drug discovery
Catalog No. 50-000-80116