Learn More
Meso Scale Discovery U-PLEX NHP TGF-B2 ASY
SDP
Supplier:  Meso Scale Discovery K156XUK2
Encompass
The Meso Scale Discovery U-PLEX NHP TGF-beta 2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of active and total TGF-beta 2 in non-human primate serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides superior sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required to perform testing of 46 samples in duplicate Turnaround time is approximately 3 5 hours This product allows for reliable reproducible quantitation of TGF-beta 2 levels
Catalog No. 50-000-81097