Learn More
Meso Scale Discovery U-PLEX NHP TGF-B2 ASY384-well
SDP
Supplier:  Meso Scale Discovery K256XUK2
Encompass
The Meso Scale Discovery U-PLEX NHP TGF-beta2 384-well Assay Kit is a multiplex immunoassay that measures active and total TGF-beta2 levels in human serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrators and controls It allows for the accurate quantification of TGF-beta2 levels from small sample volumes This assay kit can help researchers investigate TGF-beta2 s role in immune function inflammation tissue remodeling cancer and more
Catalog No. 50-000-81773
Only null left
Add to Cart
Request a Quote