Learn More
Meso Scale Discovery U-PLEX NHP TGF-B3 ASY
SDP
Supplier:  Meso Scale Discovery K156XVK1
Encompass
The Meso Scale Discovery U-PLEX NHP TGF-beta3 Assay is a multiplex immunoassay kit for measuring active and total TGF-beta3 in non-human primate samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs This assay reliably quantifies TGF-beta3 a key cytokine in immune regulation and tissue repair with minimal hands-on time making it ideal for preclinical immunology and biomarker research in NHP models
Catalog No. 50-000-80230