Learn More
Meso Scale Discovery U-PLEX NHP TGF-B3 ASY384-well
SDP
Supplier:  Meso Scale Discovery K256XVK2
Encompass
The Meso Scale Discovery U-PLEX NHP TGF beta 3 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human mouse and rat TGF beta 3 in cell culture supernatants serum plasma and other biological matrices The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and diluent The assay has a typical sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-10 000 pg/mL This multiplex assay allows high-throughput quantification of TGF beta 3 with low sample volume requirements
Catalog No. 50-000-78428
Only null left
Add to Cart
Request a Quote