Learn More
Meso Scale Discovery U-PLEX NHP TNF- ASY384-well
SDP
Supplier:  Meso Scale Discovery K256UCK2
Encompass
The Meso Scale Discovery U-PLEX NHP TNF-alpha 384-well Assay Kit is a multiplex immunoassay that measures human TNF-alpha levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrators and controls It allows high-throughput screening and only requires a small sample volume of 25 L The assay has a typical lower limit of detection of 0 04 pg/mL and an upper limit of quantitation of 10 000 pg/mL for TNF-alpha
Catalog No. 50-000-81224
Only null left
Add to Cart
Request a Quote