Learn More
Meso Scale Discovery U-PLEX NHP TNF-B ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K156UUK4
Encompass
The Meso Scale Discovery U-PLEX NHP TNF-beta Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of TNF-beta in non-human primate serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and can perform up to 70 tests across a testing range of 10-10 000 pg/mL It includes a 96-well plate pre-coated with capture antibodies detection antibodies and an MSD GOLD SULFO-TAG label The kit offers a streamlined and rapid protocol that takes around 3 hours to complete
Catalog No. 50-000-79745
Add to Cart
Request a Quote