Learn More
Meso Scale Discovery U-PLEX NHP TPO ASY
SDP
Supplier:  Meso Scale Discovery K156VKK1
Encompass
The Meso Scale Discovery U-PLEX NHP TPO Assay Kit is a multiplex immunoassay that measures thyroid peroxidase autoantibodies (TPO) in non-human primate serum plasma and other biological samples This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls Sample incubation time is just 2 hours The kit is for research use only and not for use in diagnostic procedures This assay allows for efficient quantitative measurement of TPO autoantibodies
Catalog No. 50-000-80087