Learn More
Meso Scale Discovery U-PLEX NHP VEGF-a ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K156UVK4
Encompass
The Meso Scale Discovery U-PLEX NHP VEGF-A Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of VEGF-A in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and broad dynamic range The assay has high specificity for VEGF-A with no cross-reactivity to other analytes It requires low sample volume and has a rapid 2 5 hour protocol The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrator It has a sensitivity of 0 31 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-80292
Only null left
Add to Cart
Request a Quote