Learn More
Meso Scale Discovery U-PLEX NHP VEGF-a ASY
SDP
Supplier:  Meso Scale Discovery K156UVK1
Encompass
The Meso Scale Discovery U-PLEX NHP VEGF-A Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of VEGF-A in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and broad dynamic range The assay has high specificity for VEGF-A with no cross-reactivity to related growth factors It requires low sample volume and provides rapid results in under 3 hours The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrators
Catalog No. 50-000-80241