Learn More
Meso Scale Discovery U-PLEX NHP YKL-40 ASY
SDP
Supplier:  Meso Scale Discovery K156VLK1
Encompass
The Meso Scale Discovery U-PLEX NHP YKL-40 Assay Kit is a multiplex immunoassay that quantitatively measures YKL-40 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls It can test up to 80 samples in duplicates and has a testing time of 3 5 hours The assay has a lower limit of detection of 0 31 ng/mL and a upper limit of 10 000 ng/mL
Catalog No. 50-000-79781