Learn More
Meso Scale Discovery U-PLEX Obesity Combo 1 rt
SDP
Supplier:  Meso Scale Discovery K15310K1
Encompass
The Meso Scale Discovery U-PLEX Obesity Combo 1 Rat Kit is a multiplex immunoassay kit that allows simultaneous measurement of leptin insulin and C-peptide in rat serum plasma and cell culture supernatants The kit contains a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The electrochemiluminescent assay takes approximately 3 5 hours to complete and has a detection range of 50-50 000 pg/mL for leptin 50-50 000 pg/mL for insulin and 6 4-6 400 pg/mL for C-peptide This assay kit can be used to study obesity and metabolic disease mechanisms in rat models
Catalog No. 50-000-81874