Learn More
Meso Scale Discovery U-PLEX Obesity Combo 2 rt
SDP
Supplier:  Meso Scale Discovery K15311K2
Encompass
The U-PLEX Obesity Combo 2 Rat Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of leptin insulin and C-peptide in rat serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies and calibrators The electrochemiluminescent immunoassay allows simultaneous measurement of the 3 analytes from a single sample in under 4 hours The kit has high sensitivity and a broad dynamic range This product would be useful for obesity and metabolic research in rat models
Catalog No. 50-000-79115
Add to Cart
Request a Quote